missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody (1/42), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63047CL1
This item is not returnable.
View return policy
Description
Cytomegalovirus Monoclonal antibody specifically detects Cytomegalovirus in Cytomegalovirus,Virus samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| Cytomegalovirus | |
| Monoclonal | |
| CoraFluor 1 | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| Partially purified extracts of strain AD169 infected HEL fibroblasts | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 1942-01-01 00:00:00 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction