missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus Antibody (1/42), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63047F
This item is not returnable.
View return policy
Description
Cytomegalovirus Monoclonal specifically detects Cytomegalovirus in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Cytomegalovirus | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 15342 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| CMV; HHV 5; HHV5; Human herpesvirus 5 | |
| Partially purified extracts of strain AD169 infected HEL fibroblasts | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| NB100-63047 is specific for human Cytomegalovirus and can be used to detect an early antigen 6-12 hours post infection. It does not react with laboratory strains of Adenovirus type 7 or other human Herpes viruses. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction