missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gB Antibody (hCMV34), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64532AF700
This item is not returnable.
View return policy
Description
Cytomegalovirus gB Monoclonal specifically detects Cytomegalovirus gB in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Cytomegalovirus gB | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| hCMV34 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55 | |
| Native | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Recognizes cytomegalovirus glycoprotein B, a viral ligand for CD209 which allows capture of viral particles by dendritic cells. This does not react with HSV, VSV or EBV. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction