missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytomegalovirus gB Late Antigen Monoclonal Antibody (HCMV37), FITC
GREENER_CHOICE

Product Code. 15275959
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15275959 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15275959 Supplier Invitrogen™ Supplier No. MA516701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Cytomegalovirus gB Late Neutralizing Antigen antibody, clone HCMV37 recognizes Cytomegalovirus glycoprotein B late neutralizing antigen. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus gB Late Antigen
Applications Immunocytochemistry
Classification Monoclonal
Clone HCMV37
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Alias CMV
Host Species Mouse
Immunogen Human CMV extracts.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.