missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gB Mouse anti-Cytomegalovirus, Clone: P1212, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13517.500ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against cytomegalovirus (CMV) gB.
- High specificity
- High reactivity
Specifications
| Cytomegalovirus gB | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μg | |
| Primary | |
| Cytomegalovirus (CMV) | |
| Liquid |
| Immunofluorescence | |
| P1212 | |
| Mouse monoclonal antibody raised against cytomegalovirus (CMV) gB. | |
| In PBS, pH 7.4. | |
| Protein G purification | |
| RUO | |
| This antibody reacts with gB of cytomegalovirus. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction