missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gH Antibody (HCMV16), DyLight 350 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20157UV
This item is not returnable.
View return policy
Description
Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunofluorescence
Specifications
| Cytomegalovirus gH | |
| Monoclonal | |
| DyLight 350 | |
| envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75 | |
| The original antibody was generated by immunization of mice with purified HCMV strain AD169. | |
| 0.1 mL | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
| ELISA, Immunofluorescence | |
| HCMV16 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3077450 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction