missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytomegalovirus US28-8His Recombinant Alpaca Monoclonal Antibody (Nb7)
Alpaca Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554340
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
Specifications
| Cytomegalovirus US28-8His | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Alpaca | |
| Metal-chelate chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| VHH |
| ELISA | |
| Nb7 | |
| Unconjugated | |
| US28 HCMVT | |
| Recombinant HCMV/HHV-5 HHRF3/US28. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction