missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cytoskeleton II Phospho-Specific Array

Product Code. 16012435
Click to view available options
Quantity:
1 Kit (2 Samples)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 16012435

Brand: Abnova™ AA0022.1Kit(2Samples)

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytoskeleton II Phospho-Specific Array includes 141 highly specific and well-characterized phosphorylation antibodies in the cytoskeletal pathway.

  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum

Specifications

Content And Storage 4°C
Detection Method Fluorometric
Quantity 1 Kit (2 Samples)
For Use With (Application) Profiling
Product Type Array/Library
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.