missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic beta-Glucosidase/GBA3 Antibody (OTI1F1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72100AF488
This item is not returnable.
View return policy
Description
Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Cytosolic beta-Glucosidase/GBA3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GBA3 | |
| Recombinant protein fragment corresponding to amino acids 1-150 and 370-469 of human Cytosolic beta-Glucosidase/GBA3 (NP_066024) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1F1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 57733.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction