missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1A1/SULT1A1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99855PCP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1A1/SULT1A1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| PerCP | |
| Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1 | |
| Antigen and protein A Affinity-purified | |
| Primary | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry-Paraffin | |
| Produced in rabbits immunized with purified, recombinant Human Cytosolic Sulfotransferase 1A1/SULT1A1 (Accession#: P50225-1; Glu 2-Leu 295) | |
| 0.1 mL | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction