Learn More
Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody (OTI4G1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71313R
Product Code. 18162213
  • € 751.00 / 0.10mL
Description
Cytosolic Sulfotransferase 1C2/SULT1C2 Monoclonal specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytosolic Sulfotransferase 1C2/SULT1C2
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
SULT1C2
Full length recombinant protein of human Cytosolic Sulfotransferase 1C2/SULT1C2 (NP_001047) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI4G1
Western Blot, Flow Cytometry, Immunohistochemistry
cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1CNo.1
Mouse
Affinity Purified
metabolism
6819.0
Store at 4°C in the dark.