missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody Pair [HRP]

Product Code. p-7183215 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18216746

Brand: Novus Biologicals™ NBP27931415PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cytosolic Sulfotransferase 1E1/SULT1E1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4
Gene ID (Entrez) 6783
Gene Symbol SULT1E1
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 56 ng of recombinant Cytosolic Sulfotransferase 1E1/SULT1E1.
  • Capture Antibody: 0.2 mg/mL of rabbit anti-Cytosolic Sulfotransferase 1E1/SULT1E1 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL rabbit anti-Cytosolic Sulfotransferase 1E1/SULT1E1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Breast Cancer, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Cytosolic Sulfotransferase 1E1/SULT1E1
    Technique 1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.