Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Product Code. 12698026 Shop All Epredia Products
Click to view available options
Type:
Head Seals
Sealed Head Rotor
Unit Size:
Each
Pack of 3
Product Code. 12698026
Brand: Epredia™ 59910019
Use these accessories with the Epredia™ Cytospin™ 4 Cytocentrifuge.
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
TRUSTED_SUSTAINABILITY
Specifications
Type Head Seals
For Use With (Equipment) Cytospin 4 Cytocentrifuge