missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYYR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99241-100ul
This item is not returnable.
View return policy
Description
CYYR1 Polyclonal antibody specifically detects CYYR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CYYR1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:400 | |
| C21orf95, cysteine and tyrosine-rich 1, cysteine and tyrosine-rich protein 1, cysteine/tyrosine-rich 1, Proline-rich domain-containing protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CYYR1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 116159 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction