missing translation for 'onlineSavingsMsg'
Learn More

D-2-Aminoadipic acid, 98%, Thermo Scientific™

Catalog Number p-4323513
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7620-28-2
C6H11NO4
161.16
OYIFNHCXNCRBQI-SCSAIBSYSA-N
d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
165627
CHEBI:37025
(2R)-2-aminohexanedioic acid
C(CC(C(=O)O)N)CC(=O)O
This item is not returnable. View return policy
7620-28-2
C6H11NO4
161.16
OYIFNHCXNCRBQI-SCSAIBSYSA-N
d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
165627
CHEBI:37025
(2R)-2-aminohexanedioic acid
C(CC(C(=O)O)N)CC(=O)O
Melting Point 186°C to 188°C
Color White
Elemental Analysis 43.60 to 45.84% C, 8.47 to 8.91% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -20.5
Formula Weight 161.16
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-2-Aminoadipic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'