missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford D-+-Arabitol
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
5g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 488-82-4
- Molecular Formula: C5H12O5
- Molecular Weight: 152.15 g/mol
Specifications
Specifications
| CAS | 488-82-4 |
| Molecular Formula | C5H12O5 |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction