Learn More
D-Aspartic Acid, TRC
Product Code. 30004218
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004218 1 g 1g
30010041 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004218 Supplier Toronto Research Chemicals Supplier No. TRCA7900201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1783-96-6
Formula Weight 133.0375
InChI Formula InChI=1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m1/s1
IUPAC Name (2R)-2-aminobutanedioic acid
Molecular Formula C4 H7 N O4
Molecular Weight (g/mol) 133.1
Percent Purity >95%
SMILES N[C@H](CC(=O)O)C(=O)O
Synonym (-)-Aspartic Acid, (R)-Aspartic Acid, D-(-)-Aspartic Acid, 2-Aminobutanedioic Acid, NSC 97922
Chemical Name or Material D-Aspartic acid
Grade HPLC
Show More Show Less
Specifications
Quantity 1 g
Shipping Condition Room Temperature