missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD41cc), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73038H
This item is not returnable.
View return policy
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| D-Dimer | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Protein A purified | |
| RUO | |
| D-dimer and high molecular weight fibrin degradation products, no cross-reactivity with fibrinogen | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA | |
| DD41cc | |
| Western Blot, ELISA, Sandwich ELISA | |
| D Dimer | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products. | |
| 0.2 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction