missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD44), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73039
This item is not returnable.
View return policy
Description
D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA.
Specifications
| D-Dimer | |
| Monoclonal | |
| Unconjugated | |
| D Dimer | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products. | |
| 0.2 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DD44 | |
| Western Blot 1:100-1:2000, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| D-dimer and high molecular weight fibrin degradation products, no cross-reactivity with fibrinogen | |
| Store at 4C. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction