Learn More
Melford D-Gluconic Acid
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Molecular Formula: C6H11O7K, MW 234.2
Specifications
Specifications
| Chemical Name or Material | D-Gluconic Acid |
| CAS | 299-27-4 |
| Molecular Formula | C6H11KO7 |
| Quantity | 1 kg |