Learn More
D-Glucose-1,2-13C2, TRC
Specifications
Specifications
| Chemical Name or Material | Alpha-D-Glucose-1,2-13C2 |
| CAS | 261728-61-4 |
| Molecular Formula | 13C2 C4 H12 O6 |
| Quantity | 10 mg |
| Synonym | α-D-Glucose-1,2-13C2, α-D-[1,2-13C2]Glucose, α-D-Glucopyranose-1,2-13C2 (9CI) |
| InChI Formula | InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1/i5+1,6+1 |
| SMILES | OC[C@H]1O[13C@H](O)[13C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2S,3R,4S,5R,6R)-6-(hydroxymethyl)(2,3-^{13}C_{2})oxane-2,3,4,5-tetrol |
| Molecular Weight (g/mol) | 182.14 |
| Formula Weight | 182.0701 |