missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucose-6-phosphate, dipotassium salt, hydrate, 95%, MP BIOMEDICALS
Shop All MP Biomedicals Products
Click to view available options
:
D-Glucose-6-phosphate, dipotassium salt, hydrate
Description
Usage Statement:
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Color | White |
| Quantity | 500 mg |
| Molecular Weight (g/mol) | 338.331 g/mol |
| Physical Form | Powder or Waxy Solid |
Safety and Handling
Product Identifier
- D-glucose-6-phosphate dipotassium salt hydrate
- Warning
- Specific target organ toxicity Category 2
- H371-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : D-glucose-6-phosphate dipotassium salt hydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction