missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-D-Glucose, MP Biomedicals™
Shop All MP Biomedicals Products

Click to view available options
Quantity:
10 g
100 g
1 kg
Unit Size:
100g
10g
1kg
Description
β-D-Glucose is combined into glucosides, disaccharides, oligosaccharides, polysaccharides (cellulose and starch), and glycogen. It is utilized as a metabolic intermediate by cells for either aerobic or anaerobic respiration.
D-Glucose exists in two cyclic forms, α-D-glucose and β-D-glucose, based on the position of the substituent at the anomeric center.
- α-D-Glucose is the monomer unit in starch
- β-D-glucose is the monomer unit in cellulose
- When one of these anomers is added to solution, it undergoes reversible epimerization to the other via an open-chain form
- During epimerization, the specific rotation of the solution changes gradually until it reaches equilibrium
- The existence of multiple binding modes of this single monosaccharide has been studied as a model scaffold to design compounds with increased probability of ligand binding
Specifications
Specifications
| Quantity | 1 kg |
| Formula Weight | 180.16 |
| Percent Purity | ≥97% |
| Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction