Learn More
D-Homocitrulline, TRC
Specifications
Specifications
| Chemical Name or Material | D-Homocitrulline |
| CAS | 121080-96-4 |
| Molecular Formula | C7H15N3O3 |
| Quantity | 250 mg |
| Synonym | D-Lysine, N6-(aminocarbonyl)- (9CI, ACI), N6-(Aminocarbonyl)-D-lysine (ACI), (R)-2-Amino-6-ureidohexanoic acid, (2R)-2-Amino-6-ureidohexanoic acid, D-Homocitrulline |
| InChI Formula | InChI=1S/C7H15N3O3/c8-5(6(11)12)3-1-2-4-10-7(9)13/h5H,1-4,8H2,(H,11,12)(H3,9,10,13)/t5-/m1/s1 |
| SMILES | N[C@H](CCCCNC(=O)N)C(=O)O |
| IUPAC Name | (2R)-2-amino-6-(carbamoylamino)hexanoic acid |
| Molecular Weight (g/mol) | 189.21 |
| Formula Weight | 189.1113 |