Learn More
beta-D-Lactose, ca 80% beta and 20% alpha
Product Code. p-152848 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Plastic bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10528132 500 g Plastic bottle 500g
10475651 2 kg Plastic bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10528132 Supplier Thermo Scientific Acros Supplier No. 412985000
Add to basket
missing translation for 'validMainframeMsg'
Beta-D-Lactose, CAS # 5965-66-2, is the beta anomer of the disaccharide lactose. | CAS: 5965-66-2 | C12H22O11 | 342.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Beta-D-Lactose is a disaccharide composed of galactose and glucose monomers
• In beta-D-Lactose, the orientation of the glycosidic linkages are primarily in beta form
Applications
• Beta-D-Lactose is useful for laboratory research on sugar metabolism in microbes and other organisms
• Beta-D-Lactose has been used in some laboratory studies as a carrier for inhalants because of its ability to form spherical agglomerates
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5965-66-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00064521
InChI Key GUBGYTABKSRVRQ-DCSYEGIMSA-N
Synonym beta-d-lactose, beta-lactose, lactose, lactose, anhydrous, fast-flo lactose, milk sugar, d-lactose, 4-o-beta-d-galactopyranosyl-beta-d-glucopyranose, fast-flo, lactose anhydrous
PubChem CID 6134
ChEBI CHEBI:36218
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
Specifications
Acidity 0.4mL 0.1N NaOH/6g max.
Absorbance Passes Test (per Eur. Pharm.)
Color White
Loss on Drying 0.5% max. (1 g, 80°C, 2 hrs)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 500 g
Specific Rotation Condition +55.15° (20°C c=10,water,NH3)
Specific Rotation On anhydrous basis: +54.4° to +55.9° (20°C, 589nm) (c=10, H2O/NH3)
Formula Weight 342.29
Percent Purity ≥78% (beta-lactose), 22% max. (alpha-lactose)
Physical Form Crystalline Powder
Chemical Name or Material β-D-Lactose