missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-1-phosphate cyclohexylammonium salt, 95%, Thermo Scientific™

Catalog Number 15440707
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
This item is not returnable. View return policy
MFCD00161485
Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt
This item is not returnable. View return policy
MFCD00161485
Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.5 mg
Solubility Information Soluble in water
Formula Weight 458.48
Percent Purity 95%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-1-phosphate cyclohexylammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'