missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7057697
Click to view available options
Quantity:
0.1 mg
0.5 mg
This item is not returnable. View return policy
MFCD18427943
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt
This item is not returnable. View return policy
MFCD18427943
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.1 mg
Formula Weight 588
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'