Learn More
D-Talose, TRC
Specifications
Specifications
| Chemical Name or Material | D-Talose |
| CAS | 2595-98-4 |
| Molecular Formula | C6 H12 O6 |
| Quantity | 250 mg |
| Synonym | D-Talose, Talose','D- (8CI), D(+)-Talose, NSC 224293 |
| InChI Formula | InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5-,6+/m1/s1 |
| SMILES | OC[C@@H](O)[C@H](O)[C@H](O)[C@H](O)C=O |
| IUPAC Name | (2S,3S,4S,5R)-2,3,4,5,6-pentahydroxyhexanal |
| Molecular Weight (g/mol) | 180.16 |
| Formula Weight | 180.0634 |