missing translation for 'onlineSavingsMsg'
Learn More

D-[UL-13C6]Glucosamine Hydrochloride, TRC

Product Code. 30003461
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003461 1 mg 1mg
30003489 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003461 Supplier Toronto Research Chemicals Supplier No. TRCG5151021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C6H14ClNO5
Molecular Weight (g/mol) 221.59
Synonym 2-Amino-2-deoxy-D-[UL-13C6]glucose Hydrochloride;
SMILES [H][13C@@]1(O)[13C@]([13CH2]O)([H])O[13C](O)([H])[13C@]([H])([NH3+])[13C@]1(O)[H].[Cl-]

Specifications

Quantity 1 mg
Chemical Name or Material D-[UL-13C6]Glucosamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.