missing translation for 'onlineSavingsMsg'
Learn More

D-Valine Benzyl Ester p-Toluenesulfonate, TRC

Product Code. 30451108
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451108 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451108 Supplier Toronto Research Chemicals Supplier No. TRCV09428010G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Valine Benzyl Ester p-Toluenesulfonate
CAS 17662-84-9
Molecular Formula C12 H17 N O2 . C7 H8 O3 S
Quantity 10 g
Synonym (R)-Benzyl 2-amino-3-methylbutanoate 4-methylbenzenesulfonate
InChI Formula InChI=1S/C12H17NO2.C7H8O3S/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10;1-6-2-4-7(5-3-6)11(8,9)10/h3-7,9,11H,8,13H2,1-2H3;2-5H,1H3,(H,8,9,10)/t11-;/m1./s1
SMILES CC(C)[C@@H](N)C(=O)OCc1ccccc1.Cc2ccc(cc2)S(=O)(=O)O
IUPAC Name benzyl (2R)-2-amino-3-methylbutanoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 379.471
Formula Weight 379.145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.