Learn More
D-Valine Benzyl Ester p-Toluenesulfonate, TRC
Product Code. 30455133
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455133 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455133 Supplier Toronto Research Chemicals Supplier No. TRCV09428025G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material D-Valine Benzyl Ester p-Toluenesulfonate
CAS 17662-84-9
Molecular Formula C12 H17 N O2 . C7 H8 O3 S
Quantity 25 g
Synonym (R)-Benzyl 2-amino-3-methylbutanoate 4-methylbenzenesulfonate
InChI Formula InChI=1S/C12H17NO2.C7H8O3S/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10;1-6-2-4-7(5-3-6)11(8,9)10/h3-7,9,11H,8,13H2,1-2H3;2-5H,1H3,(H,8,9,10)/t11-;/m1./s1
SMILES CC(C)[C@@H](N)C(=O)OCc1ccccc1.Cc2ccc(cc2)S(=O)(=O)O
IUPAC Name benzyl (2R)-2-amino-3-methylbutanoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 379.471
Formula Weight 379.145