missing translation for 'onlineSavingsMsg'
Learn More

D2-40 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ D2-40 Mouse Monoclonal Antibody.

Brand:  Epredia MS-1940-P

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 15260655

  • € 790.00 / 1mL
Estimated Shipment: 16-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

D2-40 is reactive to a 40kDa O-linked sialoglycoprotein in lymphatic endothelium, fetal testis, seminomas and dysgerminomas. D2-40 is useful in demonstrating lymphatic invasion by primary tumors and in vascular lesions. Publications suggest that D2-40 positivity in lymphovascular invasion was a more prognostic predictor of breast cancer than blood vessel invasion2. It is also considered useful in differential diagnosis of epithelioid malignant mesothelioma vs adenocarcinoma1.

Host Species: Mouse
Clone: D2-40
Isotype: IgG1
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: O-linked sialoglycoprotein
Molecular Weight: 40kDa
Positive Control: Breast carcinomas, mesothelioma and colon carcinoma
Cellular Localization: Membranous/Cytoplasmic
Recommended for: Immunohistochemistry
Specifications

Specifications

Immunohistochemistry
Monoclonal
Unconjugated
Mouse
40kDa.
1 mL
Primary
Store vial at 4°C. When stored at 2-8°C, this antibody is stable for 24 months..
IgG1
Membranous/Cytoplasmic.
D2-40.
Antibody purified from tissue culture supernatant. Prepared in 10mM PBS, pH 7.4, with 0.2% BSA and <0.1% sodium azide..
O-linked sialoglycoprotein.
Breast carcinomas, mesothelioma and colon carcinoma.
RUO
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers