Learn More
Thermo Scientific™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs.
Brand:  Thermo Scientific™ D3-MT2D
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11902975
  • € 98.75 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs. These microwell and wall combs are heavy-duty, one-piece units for use with the D3-14, A3-1 and A6 wide gel electrophoresis systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. D3-MTC, -MTD, -MT2C and -MT2D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifications
Specifications
Microwell comb
Comb
1.5 mm
Owl™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System
50 (2X)