missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (97A1015), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56725AF532
This item is not returnable.
View return policy
Description
D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| D4-GDI/RhoGDI2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ARHGDIB | |
| A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human D4-GDI/RhoGDI2 was used as the immunogen. | |
| Primary | |
| In anti-Fas treated Jurkat, an approx. 23 kDa band should be observed. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 97A1015 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| ARHGDIB Rho GDP dissociation inhibitor (GDI) beta, D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
| Mouse | |
| 0.1 mL | |
| 397 | |
| Human, Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction