missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (97A1015), DyLight 755 - (cleavage specific), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56725IR
This item is not returnable.
View return policy
Description
D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| D4-GDI/RhoGDI2 | |
| Monoclonal | |
| DyLight 755 | |
| P52566 | |
| ARHGDIB | |
| A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human D4-GDI/RhoGDI2 was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| In anti-Fas treated Jurkat, an approx. 23 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 97A1015 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 397 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction