missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DAB1 Antibody - C-terminus, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30795-50ug
This item is not returnable.
View return policy
Description
DAB1 Polyclonal antibody specifically detects DAB1 in Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DAB1 | |
| Polyclonal | |
| Western Blot : 1 μg/mL, Immunoprecipitation : 5 μg/mL | |
| disabled (Drosophila) homolog 1, disabled homolog 1, disabled homolog 1 (Drosophila) | |
| Recombinant protein corresponding to amino acids 399-555 of mouse disabled coupled to a GST fusion protein. | |
| 50 μg | |
| Growth and Development, Neuroscience, Phospho Specific, Signal Transduction | |
| 1600 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction