missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DAB1 Antibody - N-terminal, Novus Biologicals™
Goat Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | DAB1 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL, Immunoprecipitation : 5-10 μg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066953
|
Novus Biologicals™
NBP3-30794-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067034
|
Novus Biologicals™
NBP3-30794-250ug |
250 μg |
€ 508.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAB1 Polyclonal antibody specifically detects DAB1 in Mouse samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| DAB1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Goat | |
| Growth and Development, Neuroscience, Phospho Specific, Signal Transduction | |
| Phosphate buffered saline | |
| 1600 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL, Immunoprecipitation : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| disabled (Drosophila) homolog 1, disabled homolog 1, disabled homolog 1 (Drosophila) | |
| Goats were immunized with a recombinant protein corresponding to amino acids 1-179 of mouse disabled coupled to a GST fusion protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title