missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585453
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Chicken (96%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
Specifications
| DAB1 | |
| Polyclonal | |
| Unconjugated | |
| DAB1 | |
| AI956902; C630028C02Rik; Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Dab, reelin signal transducer, homolog 1 (Drosophila); Dab1; DAB1, reelin adaptor protein; disabled 1; disabled homolog 1; RP23-356E21.1; scm; scr; scrambler; yot; yotari | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13131, 1600, 266729 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 1.84 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O75553, P97318, Q8CJH2 | |
| DAB1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Dab1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction