missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran Etexilate-d11, TRC

Product Code. 30360852
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360852 25 mg 25mg
30351732 2.5 mg 2.50mg
30368257 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360852 Supplier Toronto Research Chemicals Supplier No. TRCD10014225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dabigatran Etexilate-d11
Molecular Formula C34H30D11N7O5
Quantity 25 mg
Synonym N-[[2-[[[4-[[[(Hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-Alanine Ethyl Ester-d11, BIBR 1048-d11, Prazaxa-d11
InChI Formula InChI=1S/C34H41N7O5/c1-4-6-7-10-21-46-34(44)39-32(35)24-12-15-26(16-13-24)37-23-30-38-27-22-25(14-17-28(27)40(30)3)33(43)41(20-18-31(42)45-5-2)29-11-8-9-19-36-29/h8-9,11-17,19,22,37H,4-7,10,18,20-21,23H2,1-3H3,(H2,35,39,44)/i1D3,4D2,6D2,7D2,10D2
SMILES O=C(C1=CC=C2C(N=C(CNC3=CC=C(C(NC(OCC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)=N)C=C3)N2C)=C1)N(CCC(OCC)=O)C4=NC=CC=C4
IUPAC Name ethyl 3-[[1-methyl-2-[[4-[N-(2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexoxycarbonyl)carbamimidoyl]anilino]methyl]benzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
Molecular Weight (g/mol) 638.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.