Learn More
Dabigatran Etexilate Mesilate, Mikromol™
Description
Dabigatran Etexilate Mesilate
Specifications
Specifications
| Chemical Name or Material | Dabigatran Etexilate Mesylate |
| CAS | 872728-81-9 |
| Analyte or Component Names | Dabigatran Etexilate Mesylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H41 N7 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | BIBR 1048MS, Dabigatran Etixilate Mesylate, Dabigatran etexilate mesylate, Pradaxa, beta-Alanine, N-[[2-[[[4-[[[(hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester, methanesulfonate (1:1), beta-Alanine, N-[[2-[[[4-[[[(hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester, monomethanesulfonate (9CI) |
| InChI Formula | InChI=1S/C34H41N7O5.CH4O3S/c1-4-6-7-10-21-46-34(44)39-32(35)24-12-15-26(16-13-24)37-23-30-38-27-22-25(14-17-28(27)40(30)3)33(43)41(20-18-31(42)45-5-2)29-11-8-9-19-36-29;1-5(2,3)4/h8-9,11-17,19,22,37H,4-7,10,18,20-21,23H2,1-3H3,(H2,35,39,44);1H3,(H,2,3,4) |
| SMILES | CCCCCCOC(=O)NC(=N)c1ccc(NCc2nc3cc(ccc3n2C)C(=O)N(CCC(=O)OCC)c4ccccn4)cc1.CS(=O)(=O)O |
| IUPAC Name | ethyl 3-[[2-[[4-(N-hexoxycarbonylcarbamimidoyl)anilino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate;methanesulfonic acid |
| Show More |