missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran etexilate mesylate, MedChemExpress

Product Code. 16480137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480137 50 mg 50mg
16460966 200 mg 200mg
16443758 10 mg 10mg
16406487 5 mg 5mg
16404826 100 mg 100mg
16401556 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16480137 Supplier Medchem Express Supplier No. HY10274A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Specifications

Chemical Name or Material Dabigatran etexilate mesylate
CAS 872728-81-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C35H45N7O8S
Quantity 50 mg
Synonym BIBR 1048MS Dabigatran etexilate methanesulfonate
Solubility Information DMSO : 50 mg/mL (69.08 mM; Need ultrasonic)
SMILES O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2.CS(=O)(O)=O
Molecular Weight (g/mol) 723.84
Formula Weight 723.84
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.