missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DACH1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518909
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Transcription factor that is involved in regulation of organogenesis. Seems to be a regulator of SIX1, SIX6 and probably SIX5. Corepression of precursor cell proliferation in myoblasts by SIX1 is switched to coactivation through recruitment of EYA3 to the SIX1-DACH1 complex. Transcriptional activation seems also to involve association of CREBBP. Seems to act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. Inhibits TGF-beta signaling through interaction with SMAD4 and NCOR1. Binds to chromatin DNA via its DACHbox-N domain.
Specifications
| DACH1 | |
| Polyclonal | |
| Unconjugated | |
| Dach1 | |
| Dac; dac homolog; DACH; Dach1; dachshund 1; dachshund 1 (Drosophila); dachshund family transcription factor 1; dachshund homolog 1; FLJ10138; OTTHUMP00000040839; RP11-512J14.1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1602 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UI36 | |
| Dach1 | |
| Synthetic peptide sequence (SPVENTPQNNECK) corresponding to the internal amino acids of DACH1 (aa 177-190). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction