missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DACH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552968
This item is not returnable.
View return policy
Description
Immunogen sequence: PKRTQSVTSP ENSHIMPHSV PGLMSPGIIP PTGLTAAAAA AAAATNAAIA EAMKVKKIKL EAMSNYHASN NQHGADSENG DMNSSVGSSD GSWDKETLPS SPSQGPQASI THPRMPGARS LPLSHPLNHL QQSHLLPNGL ELP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
Transcription factor that is involved in regulation of organogenesis. Seems to be a regulator of SIX1, SIX6 and probably SIX5. Corepression of precursor cell proliferation in myoblasts by SIX1 is switched to coactivation through recruitment of EYA3 to the SIX1-DACH1 complex. Transcriptional activation seems also to involve association of CREBBP. Seems to act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. Inhibits TGF-beta signaling through interaction with SMAD4 and NCOR1. Binds to chromatin DNA via its DACHbox-N domain.
Specifications
| DACH1 | |
| Polyclonal | |
| Unconjugated | |
| Dach1 | |
| Dac; dac homolog; DACH; Dach1; dachshund 1; dachshund 1 (Drosophila); dachshund family transcription factor 1; dachshund homolog 1; FLJ10138; OTTHUMP00000040839; RP11-512J14.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UI36 | |
| Dach1 | |
| Recombinant protein corresponding to Human DACH1. Recombinant protein control fragment (Product #RP-89667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction