missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DACH2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99137V
This item is not returnable.
View return policy
Description
DACH2 Polyclonal antibody specifically detects DACH2 in Human samples. It is validated for Western Blot
Specifications
| DACH2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 117154 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| Dach2, dachshund homolog 2, dachshund homolog 2 (Drosophila), FLJ31391, MGC138545 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DACH2. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction