missing translation for 'onlineSavingsMsg'
Learn More

Dactolisib, MedChemExpress

Product Code. 16461916
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16461916

Brand: Medchem Express HY50673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dactolisib (BEZ235) is an orally active and dual pan-class I PI3K and mTOR kinase inhibitor with IC50s of 4 nM/5 nM/7 nM/75 nM, and 20.7 nM for p110α/p110γ/p110δ/p110β and mTOR, respectively. Dactolisib (BEZ235) inhibits both mTORC1 and mTORC2.

Dactolisib (BEZ235) is an orally active and dual pan-class I PI3K and mTOR kinase inhibitor with IC50s of 4 nM/5 nM/7 nM/75 nM, and 20.7 nM for p110α/p110γ/p110δ/p110β and mTOR, respectively. Dactolisib (BEZ235) inhibits both mTORC1 and mTORC2.

Specifications

Chemical Name or Material Dactolisib
CAS 915019-65-7
Color White
Purity Grade Notes Research
Molecular Formula C30H23N5O
Quantity 500 mg
Synonym BEZ235 NVP-BEZ235
Solubility Information 5% TFA : 8.33 mg/mL (17.74 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 5 mg/mL (10.65 mM; ultrasonic and warming and heat to 60°C) ∣DMF : 2 mg/mL (4.26 mM; Need ultrasonic)
SMILES CN(C1=C2C3=CC(C4=CC5=CC=CC=C5N=C4)=CC=C3N=C1)C(N2C6=CC=C(C=C6)C(C)(C#N)C)=O
Molecular Weight (g/mol) 469.54
Formula Weight 469.54
Percent Purity 96.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.