missing translation for 'onlineSavingsMsg'
Learn More

DAGLA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

€ 280.00 - € 624.00

Specifications

Antigen DAGLA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18471442
View Documents
Novus Biologicals
NBP2-31856-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18141282
View Documents
Novus Biologicals
NBP2-31856
0.1 mL
€ 624.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DAGLA Polyclonal specifically detects DAGLA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DAGLA
Polyclonal
Rabbit
Cellular Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
C11orf11, DAGL(ALPHA), DAGLALPHA, DGL-alpha, diacylglycerol lipase, alpha, EC 3.1.1.-, KIAA0659, Neural stem cell-derived dendrite regulator, NSDDRchromosome 11 open reading frame 11, sn1-specific diacylglycerol lipase alpha
DAGLA
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q9Y4D2
747
This antibody was developed against a recombinant protein corresponding to amino acids: QEEPTYFAIWGDNKAFNEVIISPAMLHEHLPYVVMEGLNKVLENYNKGKTALLSAAKVMVSPTEVDLTPELIFQQQPL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.