missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAK Polyclonal Antibody
GREENER_CHOICE

Product Code. 16392245
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16392245 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16392245 Supplier Invitrogen™ Supplier No. PA585310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (83%), Rat (83%), Chimpanzee (100%), Bovine (89%). Positive Control: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene TKFC
Gene Accession No. Q3LXA3
Gene Alias ATP-dependent dihydroxyacetone kinase; BC021917; bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); Dak; DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; dihydroxyacetone kinase 2 homolog (S. cerevisiae); DKFZP586B1621; DKFZP586B1621 protein; FAD-AMP lyase (cyclic FMN forming); FAD-AMP lyase (cyclizing); FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; FMN cyclase; Glycerone kinase; MGC5621; NET45; RGD1311026; testis tissue sperm-binding protein Li 84P; TKFC; Triokinase; triokinase and FMN cyclase; triokinase, FMN cyclase; triokinase/FMN cyclase; Triose kinase; zgc:153296
Gene Symbols TKFC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DAK. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.