Learn More
Memmert™ DAkkS Factory Calibration for SN160plus
DAkkS factory calibration for a freely selectable temperature for Memmert Sterilizers.
Brand: Memmert™ SN160plusE39696
Additional Details : Weight : 0.01200kg
Description
- For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
SN160plus Sterilizer | |
DAkkS Factory Calibration |