missing translation for 'onlineSavingsMsg'
Learn More

Danoprevir, MedChemExpress

Product Code. 16493728
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493728

Brand: Medchem Express HY10238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Danoprevir (ITMN-191) is an orally active NS3/4A protease inhibitor for hepatitis C virus (HCV) with an IC50 of 0.29 nM and is selective for NS3/4A over a panel of 53 proteases (IC50 higher than 10 μM). Danoprevir (ITMN-191) inhibits HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s=0.2-0.4 nM) as well as 2b and 3a (IC50s=1.6, 3.5 nM). Danoprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 0.05 μM.

Danoprevir (ITMN-191) is an orally active NS3/4A protease inhibitor for hepatitis C virus (HCV) with an IC50 of 0.29 nM and is selective for NS3/4A over a panel of 53 proteases (IC50 higher than 10 μM). Danoprevir (ITMN-191) inhibits HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s=0.2-0.4 nM) as well as 2b and 3a (IC50s=1.6, 3.5 nM). Danoprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 0.05 μM.

Specifications

Chemical Name or Material Danoprevir
CAS 850876-88-9
Color White
Purity Grade Notes Research
Molecular Formula C35H46FN5O9S
Quantity 10 mg
Synonym ITMN-191 R7227 RO5190591 RG7227
Solubility Information DMSO : ≥ 100 mg/mL (136.64 mM)
SMILES O=S(NC([C@@]1(NC([C@]2([H])C[C@@H](OC(N3CC4=C(C3)C=CC=C4F)=O)CN2C5=O)=O)C[C@]1(/C=C/CCCCC[C@@H]5NC(OC(C)(C)C)=O)[H])=O)(C6CC6)=O
Molecular Weight (g/mol) 731.83
Formula Weight 731.83
Percent Purity 95.65%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.