Learn More
UBE2M/Ubc12 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00093PCP
This item is not returnable.
View return policy
Description
UBE2M/Ubc12 Polyclonal antibody specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Western Blot | |
| PerCP | |
| Antigen and protein A Affinity-purified | |
| IgG |
| Polyclonal | |
| PBS | |
| Purified |