Learn More
UBE2M/Ubc12 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00093PCP
Product Code. 18621906
  • € 407.00 / 0.10mL
Description
UBE2M/Ubc12 Polyclonal antibody specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
PerCP
Antigen and protein A Affinity-purified